Anti-PPP2R3A Goat Polyclonal Antibody

Supplier: Biorbyt
BIRBORB20185-100EA 372 CHF
BIRBORB20185-100
Anti-PPP2R3A Goat Polyclonal Antibody
Antibodies
ELISA: 1:1000; Western Blot: 1 ?g/ml

Type: Primary
Antigen: PPP2R3A (protein phosphatase 2, regulatory subunit B'', alpha)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR