Anti-KCNQ5 Goat Polyclonal Antibody

Supplier: Biorbyt
BIRBORB20181-100EA 372 CHF
BIRBORB20181-100
Anti-KCNQ5 Goat Polyclonal Antibody
Antibodies
ELISA: 1:64000; Western Blot: 3 ?g/ml

Type: Primary
Antigen: KCNQ5 (potassium voltage-gated channel, KQT-like subfamily, member 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR