Anti-SLC10A2 Goat Polyclonal Antibody

Supplier: Biorbyt
BIRBORB20178-100EA 559 CHF
BIRBORB20178-100
Anti-SLC10A2 Goat Polyclonal Antibody
Antibodies
ELISA: 1:16000; Western Blot: 1 ?g/ml

Type: Primary
Antigen: SLC10A2 (solute carrier family 10 (sodium/bile acid cotransporter), member 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR