Anti-CHRFAM7A Goat Polyclonal Antibody

Supplier: Biorbyt
BIRBORB20176-100EA 372 CHF
BIRBORB20176-100
Anti-CHRFAM7A Goat Polyclonal Antibody
Antibodies
ELISA: 1:1000; Western Blot: 1 ?g/ml

Type: Primary
Antigen: CHRFAM7A (CHRNA7 (cholinergic receptor, nicotinic, alpha 7, exons 5-10) and FAM7A (family with sequence similarity 7A, exons A-E) fusion)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR