Anti-MC5R Goat Polyclonal Antibody

Supplier: Biorbyt
BIRBORB20150-100EA 559 CHF
BIRBORB20150-100
Anti-MC5R Goat Polyclonal Antibody
Antibodies
ELISA: 1:8000; Western Blot: 0.3-1 ?g/ml

Type: Primary
Antigen: MC5R (melanocortin 5 receptor)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR