Anti-COG7 Goat Polyclonal Antibody

Lieferant: Biorbyt
BIRBORB20034-100EA 559 CHF
BIRBORB20034-100
Anti-COG7 Goat Polyclonal Antibody
Antikörper
ELISA: 1:32000; Western Blot: 1 ?g/ml

Type: Primary
Antigen: COG7 (component of oligomeric golgi complex 7)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR