Anti-ANGPTL3 Goat Polyclonal Antibody

Supplier: Biorbyt
BIRBORB20008-100EA 372 CHF
BIRBORB20008-100
Anti-ANGPTL3 Goat Polyclonal Antibody
Antibodies
ELISA: 1:8000; Western Blot: 0.3 ?g/ml

Type: Primary
Antigen: ANGPTL3 (angiopoietin-like 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR