Anti-CDH23 Goat Polyclonal Antibody

Supplier: Biorbyt
BIRBORB19984-100EA 559 CHF
BIRBORB19984-100
Anti-CDH23 Goat Polyclonal Antibody
Antibodies
Type: Primary
Antigen: CDH23 (cadherin-related 23)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR