Anti-ABCG4 Goat Polyclonal Antibody

Supplier: Biorbyt
BIRBORB19952-100EA 372 CHF
BIRBORB19952-100
Anti-ABCG4 Goat Polyclonal Antibody
Antibodies
ELISA: 1:8000; Western Blot: 1 ?g/ml

Type: Primary
Antigen: ABCG4 (ATP-binding cassette, sub-family G (WHITE), member 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR