Anti-RAMP1 Goat Polyclonal Antibody

Lieferant: Biorbyt
BIRBORB19922-100EA 559 CHF
BIRBORB19922-100
Anti-RAMP1 Goat Polyclonal Antibody
Antikörper
ELISA: 1:64000; Western Blot: 0.3-1 ?g/ml

Type: Primary
Antigen: RAMP1 (receptor (G protein-coupled) activity modifying protein 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR