Anti-ABCA5 Goat Polyclonal Antibody

Lieferant: Biorbyt
BIRBORB19916-100EA 0 CHF
BIRBORB19916-100
Anti-ABCA5 Goat Polyclonal Antibody
Antikörper
ELISA: 1:1000; Western Blot: 1 ?g/ml

Type: Primary
Antigen: ABCA5 (ATP-binding cassette, sub-family A (ABC1), member 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR