Anti-ABCD3 Goat Polyclonal Antibody

Supplier: Biorbyt
BIRBORB19913-100EA 559 CHF
BIRBORB19913-100
Anti-ABCD3 Goat Polyclonal Antibody
Antibodies
ELISA: 1:16000

Type: Primary
Antigen: ABCD3 (ATP-binding cassette, sub-family D (ALD), member 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR