Anti-ETV1 Goat Polyclonal Antibody

Supplier: Biorbyt
BIRBORB19829-100EA 372 CHF
BIRBORB19829-100
Anti-ETV1 Goat Polyclonal Antibody
Antibodies
ELISA: 1:8000; Western Blot: 1 ?g/ml

Type: Primary
Antigen: ETV1 (ets variant 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR