Anti-PNPLA3 Goat Polyclonal Antibody

Supplier: Biorbyt
BIRBORB19804-100EA 559 CHF
BIRBORB19804-100
Anti-PNPLA3 Goat Polyclonal Antibody
Antibodies
ELISA: 1:128000; Western Blot: 0.05-0.2 ?g/ml

Type: Primary
Antigen: PNPLA3 (patatin-like phospholipase domain containing 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR