Anti-PNPLA2 Goat Polyclonal Antibody

Supplier: Biorbyt
BIRBORB19789-100EA 559 CHF
BIRBORB19789-100
Anti-PNPLA2 Goat Polyclonal Antibody
Antibodies
ELISA: 1:8000; Western Blot: 0.2-0.6 ?g/ml

Type: Primary
Antigen: PNPLA2 (patatin-like phospholipase domain containing 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR