Anti-MARCH4 Goat Polyclonal Antibody

Supplier: Biorbyt
BIRBORB19773-100EA 372 CHF
BIRBORB19773-100
Anti-MARCH4 Goat Polyclonal Antibody
Antibodies
ELISA: 1:4000; Western Blot: 1 ?g/ml

Type: Primary
Antigen: MARCH4 (membrane-associated ring finger (C3HC4) 4, E3 ubiquitin protein ligase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR