Anti-CHEK2 Rabbit Polyclonal Antibody
Lieferant: Biorbyt
BIRBORB18109-100EA 616 CHF
BIRBORB18109-100
Anti-CHEK2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: CHEK2 (checkpoint kinase 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human, Mouse, Rat