Anti-MMP16 Rabbit Polyclonal Antibody
Lieferant: Biorbyt
BIRBORB18099-100EA 616 CHF
BIRBORB18099-100
Anti-MMP16 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: MMP16 (matrix metallopeptidase 16 (membrane-inserted))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human, Mouse, Rat