Anti-FAS Rabbit Polyclonal Antibody

Lieferant: Biorbyt
BIRBORB18095-100EA 0 CHF
BIRBORB18095-100
Anti-FAS Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: FAS (Fas cell surface death receptor)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR