Anti-PTPN11 Rabbit Polyclonal Antibody
Lieferant: Biorbyt
BIRBORB18090-100EA 616 CHF
BIRBORB18090-100
Anti-PTPN11 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: PTPN11 (protein tyrosine phosphatase, non-receptor type 11)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human, Mouse, Rat