Anti-LMNA Rabbit Polyclonal Antibody
Lieferant: Biorbyt
BIRBORB18081-100EA 616 CHF
BIRBORB18081-100
Anti-LMNA Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: LMNA (lamin A/C)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human, Mouse, Rat