Anti-SKP2 Rabbit Polyclonal Antibody
Lieferant: Biorbyt
BIRBORB18080-100EA 616 CHF
BIRBORB18080-100
Anti-SKP2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: SKP2 (S-phase kinase-associated protein 2, E3 ubiquitin protein ligase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human, Mouse, Rat