Anti-TIMP3 Rabbit Polyclonal Antibody

Lieferant: Biorbyt
BIRBORB18074-100EA 616 CHF
BIRBORB18074-100
Anti-TIMP3 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: TIMP3 (TIMP metallopeptidase inhibitor 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR