Anti-SOCS1 Rabbit Polyclonal Antibody
Lieferant: Biorbyt
BIRBORB18071-100EA 616 CHF
BIRBORB18071-100
Anti-SOCS1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: SOCS1 (suppressor of cytokine signaling 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human, Mouse, Rat