Anti-SCG3 Rabbit Polyclonal Antibody

Supplier: Biorbyt
BIRBORB18068-100EA 589 CHF
BIRBORB18068-100
Anti-SCG3 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: SCG3 (secretogranin III)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR