Anti-SLC16A1 Rabbit Polyclonal Antibody (FITC (Fluorescein))

Lieferant: Biorbyt
BIRBORB15960-100EA 0 CHF
BIRBORB15960-100
Anti-SLC16A1 Rabbit Polyclonal Antibody (FITC (Fluorescein))
Antikörper
Immunofluorescence: 1:50-200 (based on 0.5 mg/ml)

Type: Primary
Antigen: SLC16A1 (solute carrier family 16 (monocarboxylate transporter), member 1)
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR