Anti-JUND Rabbit Polyclonal Antibody (FITC (Fluorescein))

Lieferant: Biorbyt
BIRBORB15887-100EA 529 CHF
BIRBORB15887-100
Anti-JUND Rabbit Polyclonal Antibody (FITC (Fluorescein))
Antikörper
Immunofluorescence: 1:50-200 (based on 0.5 mg/ml)

Type: Primary
Antigen: JUND (jun D proto-oncogene)
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR