Anti-SNX25 Rabbit Polyclonal Antibody (FITC (Fluorescein))
Lieferant: Biorbyt
BIRBORB7227-100EA 0 CHF
BIRBORB7227-100
Anti-SNX25 Rabbit Polyclonal Antibody (FITC (Fluorescein))
Antikörper
Immunofluorescence: 1:50-200 (based on 0.5 mg/ml)
Type: Primary
Antigen: SNX25 (sorting nexin 25)
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat