Anti-ZNRF4 Mouse Polyclonal Antibody

ABNOH00148066-B01EA 405 CHF
ABNOH00148066-B01
Anti-ZNRF4 Mouse Polyclonal Antibody
Type: Primary
Antigen: ZNRF4 (zinc and ring finger 4)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR