Anti-ZNF563 Mouse Polyclonal Antibody

Lieferant: Abnova
ABNOH00147837-B01PEA 473 CHF
ABNOH00147837-B01P
Anti-ZNF563 Mouse Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ZNF563
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR