Anti-C18ORF23 Mouse Monoclonal Antibody [clone: 3A1]

Lieferant: Abnova
ABNOH00147341-M01EA 473 CHF
ABNOH00147341-M01
Anti-C18ORF23 Mouse Monoclonal Antibody [clone: 3A1]
Antikörper
Type: Primary
Antigen: C18ORF23
Clonality: Monoclonal
Clone: 3A1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR