Anti-HEYL Mouse Monoclonal Antibody [clone: 4A1]

Lieferant: Abnova
ABNOH00026508-M01EA 473 CHF
ABNOH00026508-M01
Anti-HEYL Mouse Monoclonal Antibody [clone: 4A1]
Antikörper
Type: Primary
Antigen: HEYL
Clonality: Monoclonal
Clone: 4A1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR