Anti-NARF Mouse Monoclonal Antibody [clone: 7D9]
Lieferant: Abnova
ABNOH00026502-M03AEA 473 CHF
ABNOH00026502-M03A
Anti-NARF Mouse Monoclonal Antibody [clone: 7D9]
Antikörper
Type: Primary
Antigen: NARF
Clonality: Monoclonal
Clone: 7D9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human