Anti-LHX6 Mouse Monoclonal Antibody [clone: 3D9]

Lieferant: Abnova
ABNOH00026468-M01EA 473 CHF
ABNOH00026468-M01
Anti-LHX6 Mouse Monoclonal Antibody [clone: 3D9]
Antikörper
Type: Primary
Antigen: LHX6
Clonality: Monoclonal
Clone: 3D9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR