Anti-GNL3 Mouse Monoclonal Antibody [clone: 4B10]

Lieferant: Abnova
ABNOH00026354-M09EA 473 CHF
ABNOH00026354-M09
Anti-GNL3 Mouse Monoclonal Antibody [clone: 4B10]
Antikörper
Type: Primary
Antigen: GNL3
Clonality: Monoclonal
Clone: 4B10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR