Anti-GNL3 Mouse Monoclonal Antibody [clone: 1G4]

Lieferant: Abnova
ABNOH00026354-M08EA 473 CHF
ABNOH00026354-M08
Anti-GNL3 Mouse Monoclonal Antibody [clone: 1G4]
Antikörper
Type: Primary
Antigen: GNL3
Clonality: Monoclonal
Clone: 1G4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR