Anti-HSPB8 Mouse Monoclonal Antibody [clone: 3C5]

Lieferant: Abnova
ABNOH00026353-M12EA 473 CHF
ABNOH00026353-M12
Anti-HSPB8 Mouse Monoclonal Antibody [clone: 3C5]
Antikörper
Type: Primary
Antigen: HSPB8
Clonality: Monoclonal
Clone: 3C5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR