Anti-HSPB8 Mouse Monoclonal Antibody [clone: 5G3]

Lieferant: Abnova
ABNOH00026353-M05EA 473 CHF
ABNOH00026353-M05
Anti-HSPB8 Mouse Monoclonal Antibody [clone: 5G3]
Antikörper
Type: Primary
Antigen: HSPB8
Clonality: Monoclonal
Clone: 5G3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR