Anti-HSPB8 Mouse Polyclonal Antibody
Lieferant: Abnova
ABNOH00026353-A01EA 310 CHF
ABNOH00026353-A01
Anti-HSPB8 Mouse Polyclonal Antibody
Antikörper
Type: Primary
Antigen: HSPB8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human