Anti-OR5H1 Mouse Polyclonal Antibody

Lieferant: Abnova
ABNOH00026341-B01EA 473 CHF
ABNOH00026341-B01
Anti-OR5H1 Mouse Polyclonal Antibody
Antikörper
Type: Primary
Antigen: OR5H1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR