Anti-GAPDHS Rabbit Polyclonal Antibody

Lieferant: Abnova
ABNOH00026330-D01PEA 456 CHF
ABNOH00026330-D01P
Anti-GAPDHS Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: GAPDHS
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR