Anti-MYCBP Mouse Monoclonal Antibody [clone: 1B12]

Lieferant: Abnova
ABNOH00026292-M13EA 456 CHF
ABNOH00026292-M13
Anti-MYCBP Mouse Monoclonal Antibody [clone: 1B12]
Antikörper
Type: Primary
Antigen: MYCBP
Clonality: Monoclonal
Clone: 1B12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR