Anti-FGF21 Mouse Monoclonal Antibody [clone: 3E8]

Lieferant: Abnova
ABNOH00026291-M05EA 473 CHF
ABNOH00026291-M05
Anti-FGF21 Mouse Monoclonal Antibody [clone: 3E8]
Antikörper
Type: Primary
Antigen: FGF21
Clonality: Monoclonal
Clone: 3E8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR