Anti-FGF21 Mouse Monoclonal Antibody [clone: 1A8]
Lieferant: Abnova
ABNOH00026291-M02EA 473 CHF
ABNOH00026291-M02
Anti-FGF21 Mouse Monoclonal Antibody [clone: 1A8]
Antikörper
Type: Primary
Antigen: FGF21
Clonality: Monoclonal
Clone: 1A8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human