Anti-NNT Mouse Monoclonal Antibody [clone: 1D6]
Lieferant: Abnova
ABNOH00023530-M01EA 473 CHF
ABNOH00023530-M01
Anti-NNT Mouse Monoclonal Antibody [clone: 1D6]
Antikörper
Type: Primary
Antigen: NNT
Clonality: Monoclonal
Clone: 1D6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human