Anti-CLCF1 Mouse Polyclonal Antibody

Lieferant: Abnova
ABNOH00023529-B03PEA 473 CHF
ABNOH00023529-B03P
Anti-CLCF1 Mouse Polyclonal Antibody
Antikörper
Type: Primary
Antigen: CLCF1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR