Anti-ZNF281 Mouse Monoclonal Antibody [clone: 1C9]
Lieferant: Abnova
ABNOH00023528-M02EA 473 CHF
ABNOH00023528-M02
Anti-ZNF281 Mouse Monoclonal Antibody [clone: 1C9]
Antikörper
Type: Primary
Antigen: ZNF281
Clonality: Monoclonal
Clone: 1C9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human