Anti-ACAP2 Mouse Monoclonal Antibody [clone: 4G3]

Lieferant: Abnova
ABNOH00023527-M01EA 473 CHF
ABNOH00023527-M01
Anti-ACAP2 Mouse Monoclonal Antibody [clone: 4G3]
Antikörper
Type: Primary
Antigen: ACAP2
Clonality: Monoclonal
Clone: 4G3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR