Anti-CABIN1 Mouse Monoclonal Antibody [clone: 2F5]

Lieferant: Abnova
ABNOH00023523-M02EA 473 CHF
ABNOH00023523-M02
Anti-CABIN1 Mouse Monoclonal Antibody [clone: 2F5]
Antikörper
Type: Primary
Antigen: CABIN1
Clonality: Monoclonal
Clone: 2F5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR