Anti-CABIN1 Mouse Polyclonal Antibody
Lieferant: Abnova
ABNOH00023523-A01EA 310 CHF
ABNOH00023523-A01
Anti-CABIN1 Mouse Polyclonal Antibody
Antikörper
Type: Primary
Antigen: CABIN1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human